Methyl-3-isochinolincarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL338559-1GEA 76.6 EUR
SIAL338559-1G
Methyl-3-isochinolincarboxylat, Sigma-Aldrich®
Methyl-3-isochinolincarboxylat
Formel: C₁₁H₉NO₂
MDL Number: MFCD00075138
CAS-Nummer: 27104-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR