2-Nitro-1-naphthaldehyd, Sigma-Aldrich®

Lieferant: Merck

SIAL555592-1GEA 137 EUR
SIAL555592-1G SIAL555592-5G
2-Nitro-1-naphthaldehyd, Sigma-Aldrich®
2-Nitro-1-naphthaldehyd
Formel: C₁₁H₇NO₃
MDL Number: MFCD03427164
CAS-Nummer: 101327-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR