2-Methoxy-4-methylpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL578525-5GEA 162 EUR
SIAL578525-5G
2-Methoxy-4-methylpyridin, Sigma-Aldrich®
2-Methoxy-4-methylpyridine
Formel: C₇H₉NO
Molekulargewicht: 123,1525 g/mol
MDL Number: MFCD06200799
CAS-Nummer: 100848-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR