4-Cyan-4'-heptylbiphenyl, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL330817-1GEA 102 EUR
SIAL330817-1G
4-Cyan-4'-heptylbiphenyl, Sigma-Aldrich®
4-Cyan-4'-heptylbiphenyl
Formel: C₂₀H₂₃N
Molekulargewicht: 277,41 g/mol
MDL Number: MFCD00074915
CAS-Nummer: 41122-71-8
EINECS: 255-229-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR