4-Methyl-5-decanol (Gemisch der Stereoisomere) erythro + threo ≥99%
B20264.06EA
26.9
EUR
B20264.06
4-Methyl-5-decanol (Gemisch der Stereoisomere) erythro + threo ≥99%
4-Methyl-5-decanol (Gemisch der Stereoisomere)
Formel:
C₁₁H₂₄O |
MDL Number:
MFCD00046700 CAS-Nummer: 213547-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...