1,2,3-Benzoltricarbonsäure Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALB4201-10GEA 133 EUR
SIALB4201-10G
1,2,3-Benzoltricarbonsäure Hydrat, Sigma-Aldrich®
1,2,3-Benzoltricarbonsäure Hydrat
Formel: C₉H₆O₆·H₂O
Molekulargewicht: 210,14 g/mol (anhy)
MDL Number: MFCD00002468
CAS-Nummer: 732304-21-1
EINECS: 209-317-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR