5-Brom-2-(tributylstannyl)pyridin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

SIAL704830-1GEA 59.4 EUR
SIAL704830-1G
5-Brom-2-(tributylstannyl)pyridin, Sigma-Aldrich®
5-Brom-2-(tributylstannyl)pyridin
Formel: C₁₇H₃₀BrNSn
MDL Number: MFCD10699138
CAS-Nummer: 611168-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR