Filters for X-plore® 2100 half masks
Lieferant: Dräger
X-plore®
DRAE6736777EA 56 EUR
DRAE6736777
Filters for X-plore® 2100 half masks
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
Half mask particle filter with protection level P100.
  • High filtration efficiency
  • Due to a large surface area low breathing resistance
  • Long life time
Zertifizierungen: NIOSH approved