HPLC columns, Acclaim™ Carbamate
Lieferant: Thermo Fisher Scientific
Acclaim™
555-7488EA 205 EUR
555-7488 555-7487 555-7484 555-7486
HPLC columns, Acclaim™ Carbamate
Chromatographiesäulen
Columns are used for the separation of carbamate pesticides.
  • Compatible with both binary and ternary mobile phase gradients
  • High efficiency
  • Low column bleed
  • Rugged column packing