Methansulfonamid

Lieferant: Apollo Scientific

APOSOR18387-500GEA 63.9 EUR
APOSOR18387-500G
Methansulfonamid
Methansulfonamid
Formel: CH₅NO₂S
Molekulargewicht: 95,13
MDL Number: MFCD00007940
CAS-Nummer: 3144-09-0
EINECS: 221-553-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR