2-Brom-5-fluorbenzamid 96%
|
Achtung
|
H62366.03EA
181
EUR
H62366.03
2-Brom-5-fluorbenzamid 96%
2-Brom-5-fluorbenzamid
Formel:
C₇H₅BrFNO Molekulargewicht: 218,02 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03094322 CAS-Nummer: 1006-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...