Zinn(II)acetylacetonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL697478-5GEA 151 EUR
SIAL697478-5G
Zinn(II)acetylacetonat, Sigma-Aldrich®
Zinn(II)acetylacetonat
Formel: C₁₀H₁₄O₄Sn
Molekulargewicht: 316,93 g/mol
MDL Number: MFCD00078005
CAS-Nummer: 16009-86-2
EINECS: 240-142-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR