Dibutylbis(2,4-pentandionato-O,O')zinn, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL520586-500MLEA 182 EUR
SIAL520586-500ML
Dibutylbis(2,4-pentandionato-O,O')zinn, Sigma-Aldrich®
Dibutylbis(2,4-pentandionato-O,O')zinn
Formel: C₁₈H₃₂O₄Sn
Molekulargewicht: 431,15 g/mol
MDL Number: MFCD00077994
CAS-Nummer: 22673-19-4
EINECS: 245-152-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR