4-(Benzyloxy)anilin Hydrochlorid

Lieferant: Molekula

Synonyme: 4-Aminophenyl benzyl ether hydrochloridep-Benzyloxyaniline hydrochloride

MOLE16990076-100GEA 96.3 EUR
MOLE16990076-100G
4-(Benzyloxy)anilin Hydrochlorid
4-(Benzyloxy)anilin Hydrochlorid
Formel: C₁₃H₁₄ClNO
MDL Number: MFCD00012995
CAS-Nummer: 51388-20-6
EINECS: 257-170-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR