4-(2-Fluor-6-nitrophenyl)morpholin ≥97%
|
Achtung
|
H63272.03EA
76.5
EUR
H63272.03
4-(2-Fluor-6-nitrophenyl)morpholin ≥97%
4-(2-Fluor-6-nitrophenyl)morpholin
Formel:
C₁₀H₁₁FN₂O₃ Molekulargewicht: 226,20 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13689260 CAS-Nummer: 1233955-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...