5,7-Dichlor-2-methyl-8-chinolinol

Lieferant: Molekula

MOLE22647958-25GEA 0 EUR
MOLE22647958-25G
5,7-Dichlor-2-methyl-8-chinolinol
5,7-Dichlor-2-methyl-8-chinolinol
Formel: C₁₀H₇Cl₂NO
Molekulargewicht: 228,0747 g/mol
MDL Number: MFCD00023984
CAS-Nummer: 72-80-0
EINECS: 200-789-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR