N-Benzyloxycarbonyl-L-alaninol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Cbz-L-Alaninol
, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester
, Z-Ala-ol
M-Clarity™ Qualitätsstufe = MQ100
SIAL543217-5GEA
136
EUR
SIAL543217-5G
N-Benzyloxycarbonyl-L-alaninol, Sigma-Aldrich®
N-Cbz-L-Alaninol
Formel:
C₁₁H₁₅NO₃ Molekulargewicht: 209,25 g/mol |
MDL Number:
MFCD00672530 CAS-Nummer: 66674-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...