6-Brompicolinamid, Sigma-Aldrich®

Lieferant: Merck

SIAL714526-1GEA 57.9 EUR
SIAL714526-1G
6-Brompicolinamid, Sigma-Aldrich®
6-Brompicolinamid
Formel: C₆H₅BrN₂O
Molekulargewicht: 201,02
MDL Number: MFCD00234154
CAS-Nummer: 25194-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR