5-Brom-N-methylpicolinamid 96%

Lieferant: Thermo Scientific
Achtung

H54947.03EA 116 EUR
H54947.03 H54947.06 H54947.MD
5-Brom-N-methylpicolinamid 96%
5-Brom-N-methylpicolinamid
Formel: C₇H₇BrN₂O
Molekulargewicht: 215,05
Schmelzpunkt: 76…77 °C
MDL Number: MFCD07357421
CAS-Nummer: 845305-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR