(R)-3-(Fmoc-amino)-3-phenylpropionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL09795-500MGEA 260 EUR
SIAL09795-500MG
(R)-3-(Fmoc-amino)-3-phenylpropionsäure, Sigma-Aldrich®
(R)-3-(Fmoc-amino)-3-phenylpropionsäure
Formel: C₂₄H₂₁NO₄
Molekulargewicht: 387,43
MDL Number: MFCD01311783
CAS-Nummer: 220498-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR