4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure ≥97%
|
Achtung
|
H52533.MDEA
95.7
EUR
H52533.MD
H52533.03
4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure ≥97%
4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure
Formel:
C₁₃H₁₉BN₂O₄ Schmelzpunkt: 100…102 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09027218 CAS-Nummer: 913835-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...