3-(Morpholino)phenylboronsäurepinakolester

MAYBCC33839DAEA 0 EUR
MAYBCC33839DA
3-(Morpholino)phenylboronsäurepinakolester
3-(Morpholino)phenylboronsäurepinakolester
Formel: C₁₆H₂₄BNO₃
Molekulargewicht: 289,1777 g/mol
MDL Number: MFCD03412097
CAS-Nummer: 852227-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR