Azocyclotin, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

45335-250MGEA 69.5 EUR
45335-250MG
Azocyclotin, Supelco®
Azocyclotin
Formel: C₂₀H₃₅N₃Sn
MDL Number: MFCD00055290
CAS-Nummer: 41083-11-8
EINECS: 255-209-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR