2,5-Bis(1-naphthyl)-1,3,4-oxadiazol, Sigma-Aldrich®

Lieferant: Merck

SIAL698202-5GEA 270 EUR
SIAL698202-5G
2,5-Bis(1-naphthyl)-1,3,4-oxadiazol, Sigma-Aldrich®
2,5-Bis(1-naphthyl)-1,3,4-oxadiazol
Formel: C₂₂H₁₄N₂O
Molekulargewicht: 322,37
MDL Number: MFCD00045582
CAS-Nummer: 905-62-4
EINECS: 212-995-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR