1-(3-Aminophenylsulfonyl)pyrrolidin

Lieferant: Thermo Scientific

MAYBMO07110DAEA 0 EUR
MAYBMO07110DA
1-(3-Aminophenylsulfonyl)pyrrolidin
1-(3-Aminophenylsulfonyl)pyrrolidin
Formel: C₁₀H₁₄N₂O₂S
MDL Number: MFCD02615287
CAS-Nummer: 91619-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR