1-(2-Methoxyphenyl)piperazin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL270067-100GEA 195 EUR
SIAL270067-100G
1-(2-Methoxyphenyl)piperazin Hydrochlorid, Sigma-Aldrich®
1-(2-Methoxyphenyl)piperazin Hydrochlorid
Formel: C₁₁H₁₇ClN₂O
Molekulargewicht: 228,72
MDL Number: MFCD00012762
CAS-Nummer: 5464-78-8
EINECS: 226-762-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR