5-Amino-2-(trifluormethyl)benzimidazol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL579483-1GEA
55
EUR
SIAL579483-1G
5-Amino-2-(trifluormethyl)benzimidazol, Sigma-Aldrich®
5-Amino-2-(trifluoromethyl)benzimidazole
Formel:
C₈H₆F₃N₃ Molekulargewicht: 201,15 g/mol |
MDL Number:
MFCD01632182 CAS-Nummer: 3671-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...