1-Methyl-4-imidazolsulfonylchlorid
Lieferant: Thermo Scientific
MAYBCC03603CBEA
0
EUR
MAYBCC03603CB
MAYBCC03603DA
1-Methyl-4-imidazolsulfonylchlorid
1-Methyl-4-imidazolsulfonylchlorid
Formel:
C₄H₅ClN₂O₂S Molekulargewicht: 180,61 g/mol |
MDL Number:
MFCD00068060 CAS-Nummer: 137049-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...