1-Acetylpyren, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL391425-10GEA 213 EUR
SIAL391425-10G
1-Acetylpyren, Sigma-Aldrich®
1-Acetylpyren
Formel: C₁₈H₁₂O
Molekulargewicht: 244,29 g/mol
MDL Number: MFCD00021663
CAS-Nummer: 3264-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR