Tributyl(4-fluorphenyl)stannan
|
Gefahr
|
APOSPC9284-1GEA
113
EUR
APOSPC9284-1G
Tributyl(4-fluorphenyl)stannan
Tributyl(4-fluorphenyl)stannan
Formel:
C₁₈H₃₁FSn |
MDL Number:
MFCD01313817 CAS-Nummer: 17151-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...