Tributyl(4-fluorphenyl)stannan

Lieferant: Thermo Scientific
Gefahr

MAYBSP00184DAEA 0 EUR
MAYBSP00184DA MAYBSP00184.10
Tributyl(4-fluorphenyl)stannan
Tributyl(4-fluorphenyl)stannan
Formel: C₁₈H₃₁FSn
MDL Number: MFCD01313817
CAS-Nummer: 17151-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR