2-Fluor-α-methyl-4-biphenylessigsäure
Lieferant: Molekula
Synonyme:
Froben
, Ansaid
, Antadys
, Flurbiprofen
MOLE26685423-1GEA
0
EUR
MOLE26685423-1G
2-Fluor-α-methyl-4-biphenylessigsäure
2-Fluor-α-methyl-4-biphenylessigsäure
Formel:
C₁₅H₁₃FO₂ Molekulargewicht: 244,265 g/mol |
MDL Number:
MFCD00079303 CAS-Nummer: 5104-49-4 EINECS: 225-827-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...