(S)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL05102-500MG-FEA 112 EUR
SIAL05102-500MG-F
(S)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(4-fluorphenyl)propionsäure
Formel: C₁₄H₁₈FNO₄
MDL Number: MFCD03427919
CAS-Nummer: 479064-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR