3-(Boc-amino)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL578843-1GEA 75 EUR
SIAL578843-1G
3-(Boc-amino)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Boc-amino)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₄
Molekulargewicht: 319,21 g/mol
MDL Number: MFCD03789256
CAS-Nummer: 330793-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR