4-Brombenzoldiazoniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL280895-5GEA 66.6 EUR
SIAL280895-5G
4-Brombenzoldiazoniumtetrafluoroborat, Sigma-Aldrich®
4-Bromobenzenediazonium tetrafluoroborate
Formel: C₆H₄BBrF₄N₂
Molekulargewicht: 270,83 g/mol
MDL Number: MFCD00011894
CAS-Nummer: 673-40-5
EINECS: 211-608-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR