N-Boc-3-bromanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL528706-1GEA 25 EUR
SIAL528706-1G
N-Boc-3-bromanilin, Sigma-Aldrich®
N-Boc-3-bromanilin
Formel: C₁₁H₁₄BrNO₂
Molekulargewicht: 272,1400 g/mol
MDL Number: MFCD01006618
CAS-Nummer: 25216-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR