N-Boc-2-iodanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL647314-1GEA 151 EUR
SIAL647314-1G SIAL647314-10G
N-Boc-2-iodanilin, Sigma-Aldrich®
N-Boc-2-iodanilin
Formel: C₁₁H₁₄INO₂
MDL Number: MFCD07369726
CAS-Nummer: 161117-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR