Natrium-p-nitrophenoxid Dihydrat

Lieferant: EHRENSTORFER GMBH

EHERC15594400EA 60.8 EUR
EHERC15594400
Natrium-p-nitrophenoxid Dihydrat
Natrium-p-nitrophenoxid Dihydrat
Formel: C₆H₄NNaO₃·2H₂O
MDL Number: MFCD00150721
CAS-Nummer: 66924-59-2
EINECS: 212-536-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR