Natrium-p-nitrophenoxid Dihydrat, Supelco®

Lieferant: Merck

36612-1G-REA 64.9 EUR
36612-1G-R
Natrium-p-nitrophenoxid Dihydrat, Supelco®
Natrium-p-nitrophenoxid Dihydrat
Formel: C₆H₄NNaO₃·2H₂O
MDL Number: MFCD00150721
CAS-Nummer: 66924-59-2
EINECS: 212-536-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR