2,4-Dinitrobenzolsulfonsäure Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL556971-25GEA
120
EUR
SIAL556971-25G
SIAL556971-5G
2,4-Dinitrobenzolsulfonsäure Hydrat, Sigma-Aldrich®
2,4-Dinitrobenzolsulfonsäure Hydrat
Formel:
C₆H₄N₂O₇S·H₂O Molekulargewicht: 248,17 g/mol (anhy) |
MDL Number:
MFCD00150961 CAS-Nummer: 89-02-1 EINECS: 201-876-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...