6-(Chlormethyl)uracil, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL183946-5GEA 110 EUR
SIAL183946-5G
6-(Chlormethyl)uracil, Sigma-Aldrich®
6-(Chlormethyl)uracil
Formel: C₅H₅ClN₂O₂
Molekulargewicht: 160,56 g/mol
MDL Number: MFCD00010095
CAS-Nummer: 18592-13-7
EINECS: 242-431-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR