Wolframdichloriddioxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL383198-10GEA
282
EUR
SIAL383198-10G
Wolframdichloriddioxid, Sigma-Aldrich®
Wolframdichloriddioxid
Formel:
Cl₂O₂W Molekulargewicht: 286,75 g/mol |
MDL Number:
MFCD00054136 CAS-Nummer: 13520-76-8 EINECS: 236-862-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...