Picolinafen
Lieferant: EHRENSTORFER GMBH
EHERC16205000EA
133
EUR
EHERC16205000
Picolinafen
Picolinafen
Formel:
C₁₉H₁₂F₄N₂O₂ |
MDL Number:
MFCD04040061 CAS-Nummer: 137641-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...