3-Morpholinobenzaldehyd
Lieferant: Thermo Scientific
MAYBCC33804DAEA
0
EUR
MAYBCC33804DA
3-Morpholinobenzaldehyd
3-Morpholinobenzaldehyd
Formel:
C₁₁H₁₃NO₂ Molekulargewicht: 191,2264 g/mol |
MDL Number:
MFCD07772811 CAS-Nummer: 446866-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...