4-Morpholinobenzaldehyd

Lieferant: Thermo Scientific

MAYBCC17404.250EA 0 EUR
MAYBCC17404.250
4-Morpholinobenzaldehyd
4-Morpholinobenzaldehyd
Formel: C₁₁H₁₃NO₂
Molekulargewicht: 191,2264 g/mol
MDL Number: MFCD00735826
CAS-Nummer: 1204-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR