N-[(Benzyloxy)carbonyl]-L-tryptophan

Lieferant: Molekula

MOLE20706773-25GEA 68 EUR
MOLE20706773-25G
N-[(Benzyloxy)carbonyl]-L-tryptophan
N(α)-Cbz-L-tryptophan
Formel: C₁₉H₁₈N₂O₄
Molekulargewicht: 338,4 g/mol
MDL Number: MFCD00065700
CAS-Nummer: 7432-21-5
EINECS: 231-074-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR