1-(4-Methoxyphenyl)cyclopropancarbonsäure 96%
ACRO170710100EA
80.8
EUR
ACRO170710100
ACRO170710500
1-(4-Methoxyphenyl)cyclopropancarbonsäure 96%
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formel:
C₁₁H₁₂O₃ Molekulargewicht: 192,2112 g/mol Schmelzpunkt: 127…132 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00019223 CAS-Nummer: 16728-01-1 EINECS: 240-788-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 127 to 132 °C |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...