Diisopentylphthalat

Lieferant: EHRENSTORFER GMBH

EHERC16173680EA 112 EUR
EHERC16173680
Diisopentylphthalat
Diisopentylphthalat
Formel: C₁₈H₂₆O₄
CAS-Nummer: 605-50-5
EINECS: 210-088-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR