4-Ethylcaprylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ300

SIALW380008-100G-KEA 854 EUR
SIALW380008-100G-K SIALW380008-25G-K
4-Ethylcaprylsäure, Sigma-Aldrich®
4-Ethylcaprylsäure
Formel: C₁₀H₂₀O₂
Molekulargewicht: 172,27 g/mol
MDL Number: MFCD00506494
CAS-Nummer: 16493-80-4
EINECS: 240-560-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR