6-Isopropylindol, Sigma-Aldrich®

Lieferant: Merck

SIAL655341-5GEA 557 EUR
SIAL655341-5G SIAL655341-1G
6-Isopropylindol, Sigma-Aldrich®
6-Isopropylindol
Formel: C₁₁H₁₃N
MDL Number: MFCD07369790
CAS-Nummer: 32996-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR